Abnova
Manufacturer Code:H00054210M01
Catalog # 89-001-297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F5 |
Immunogen | TREM1 (AAH17773.1 21 a.a. ~ 234 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant TREM1.
For Research Use Only
Protein Aliases: CD354; TREM-1; Triggering receptor expressed on monocytes 1; Triggering receptor expressed on myeloid cells 1
Gene Aliases: CD354; TREM-1; TREM1
UniProt ID: (Human) Q9NP99
Entrez Gene ID: (Human) 54210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.