Abnova
Manufacturer Code:H00000706D01P
Catalog # 89-017-458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TSPO (AAH01110.1 1 a.a. ~ 169 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human TSPO protein.
For Research Use Only
Protein Aliases: benzodiazepine peripheral binding site; BZRP; DBI; IBP; MBR; mDRC; Mitochondrial benzodiazepine receptor; PBR; Peripheral-type benzodiazepine receptor; pk18; PKBS; PTBR; Translocator protein; translocator protein (18kDa)
Gene Aliases: BPBS; BZRP; DBI; IBP; MBR; mDRC; PBR; PBS; pk18; PKBS; PTBR; TSPO
UniProt ID: (Human) P30536
Entrez Gene ID: (Human) 706
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.