Abnova
Manufacturer Code:H00009804M01
Catalog # 89-014-244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F3 |
Immunogen | TOMM20 (AAH66335 1 a.a. ~ 145 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant TOMM20.
For Research Use Only
Protein Aliases: KIAA0016; MAS20; MGC117367; Mitochondrial 20 kDa outer membrane protein; Mitochondrial import receptor subunit TOM20 homolog; MOM19; Outer mitochondrial membrane receptor Tom20; TOM20; translocase of outer mitochondrial membrane 20 homolog type II
Gene Aliases: KIAA0016; MAS20; MOM19; TOM20; TOMM20
UniProt ID: (Human) Q15388
Entrez Gene ID: (Human) 9804
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.