Proteintech
Manufacturer Code:602131IG150UL
Catalog # 50-173-6316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A10C2 |
Immunogen | transgelin/SM22 Fusion Protein Ag0764 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa actin-binding protein; Actin-associated protein p27; Protein WS3-10; SM22 SM22 alpha SM22a SM22-a SM22alpha Smooth muscle protein 22 alpha TAGLN TAGLN1 transgelin transgelin/SM22 alpha; SM22-alpha; Sm22alpha; smooth muscle 22 protein; Smooth muscle protein 22-alpha; Transgelin; transgelin variant 2
Gene Aliases: SM22; SM22A; SMCC; TAGLN; TAGLN1; WS3-10; Ws310
UniProt ID: (Human) Q01995, (Rat) P31232, (Mouse) P37804
Entrez Gene ID: (Human) 6876, (Pig) 397021, (Rat) 25123, (Mouse) 21345
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.