Abnova
Manufacturer Code:H00007041B01
Catalog # 89-006-349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | TGFB1I1 (NP_057011.2 1 a.a. ~ 444 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human TGFB1I1 protein.
For Research Use Only
Protein Aliases: Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; Androgen receptor-associated protein of 55 kDa; ARA55; HIC-5; HIC5; Hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; Transforming growth factor beta-1-induced transcript 1 protein; TSC-5
Gene Aliases: ARA55; HIC-5; HIC5; TGFB1I1; TSC-5
UniProt ID: (Human) O43294
Entrez Gene ID: (Human) 7041
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.