Abnova
Manufacturer Code:H00007023M03
Catalog # 89-001-011
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7A10 |
Immunogen | TFAP4 (NP_003214 93 a.a. ~ 192 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant TFAP4.
For Research Use Only
Protein Aliases: Activating enhancer-binding protein 4; AP-4; bHLHc41; Class C basic helix-loop-helix protein 41; Transcription factor AP-4; transcription factor AP-4 (activating enhancer-binding protein 4)
Gene Aliases: AP-4; BHLHC41; TFAP4
UniProt ID: (Human) Q01664
Entrez Gene ID: (Human) 7023
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.