Abnova
Manufacturer Code:H00083741B01
Catalog # 89-008-297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | TFAP2D (NP_758438.1 1 a.a. ~ 452 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human TFAP2D protein.
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 beta-like 1; Activating enhancer-binding protein 2-delta; AP-2 like; AP2-delta; TFAP2BL1; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); Transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta
Gene Aliases: TFAP2BL1; TFAP2D
UniProt ID: (Human) Q7Z6R9
Entrez Gene ID: (Human) 83741
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.