Abnova
Manufacturer Code:H00007021M02
Catalog # 89-001-010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F6 |
Immunogen | TFAP2B (NP_003212 73 a.a. ~ 182 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant TFAP2B.
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 beta; Activating enhancer-binding protein 2-beta; AP-2B; AP2-B; AP2-beta; MGC21381; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); Transcription factor AP-2-beta
Gene Aliases: AP-2B; AP2-B; TFAP2B
UniProt ID: (Human) Q92481
Entrez Gene ID: (Human) 7021
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.