Abnova
Manufacturer Code:H00006934M06
Catalog # 89-001-007
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A4 |
Immunogen | TCF7L2 (NP_110383 490 a.a. ~ 596 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant TCF7L2.
For Research Use Only
Protein Aliases: HMG box transcription factor 4; hTCF-4; T-cell factor 4; T-cell factor-4 variant A; T-cell factor-4 variant B; T-cell factor-4 variant C; T-cell factor-4 variant D; T-cell factor-4 variant E; T-cell factor-4 variant F; T-cell factor-4 variant G; T-cell factor-4 variant H; T-cell factor-4 variant I; T-cell factor-4 variant J; T-cell factor-4 variant K; T-cell factor-4 variant L; T-cell factor-4 variant M; T-cell factor-4 variant X2; T-cell-specific transcription factor 4; TCF-4; TCF4; Transcription factor 7-like 2; transcription factor 7-like 2 (T-cell specific, HMG-box)
Gene Aliases: TCF-4; TCF4; TCF7L2
UniProt ID: (Human) Q9NQB0
Entrez Gene ID: (Human) 6934
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.