Abnova
Manufacturer Code:H00114609B01
Catalog # 89-008-461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | TIRAP (AAH32474.1 1 a.a. ~ 256 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human TIRAP protein.
For Research Use Only
Protein Aliases: adapter protein wyatt; Adaptor protein Wyatt; FLJ42305; Mal; MyD88 adapter-like protein; MyD88-2; TIR domain-containing adapter protein; toll-interleukin 1 receptor (TIR) domain containing adaptor protein; Toll-like receptor adaptor protein; Toll/interleukin-1 receptor domain-containing adapter protein; wyatt
Gene Aliases: BACTS1; MAL; MyD88-2; TIRAP; wyatt
UniProt ID: (Human) P58753
Entrez Gene ID: (Human) 114609
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.