Abnova
Manufacturer Code:H00007273M02
Catalog # 89-001-035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B3 |
Immunogen | TTN (AAH58824 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal
For Research Use Only
Protein Aliases: CMD1G; CMH9; CMPD4; CONNECTIN; DKFZp451N061; EOMFC; FLJ26020; FLJ26409; FLJ32040; FLJ34413; FLJ39564; FLJ43066; HMERF; LGMD2J; Rhabdomyosarcoma antigen MU-RMS-40.14; Titin; TMD
Gene Aliases: CMD1G; CMH9; CMPD4; EOMFC; HMERF; LGMD2J; MYLK5; TMD; TTN
UniProt ID: (Human) Q8WZ42
Entrez Gene ID: (Human) 7273
Molecular Function: cell adhesion molecule extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.