Abnova
Manufacturer Code:PAB7339
Catalog # 89-133-162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to zebrafish thrb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of thrb.
For Research Use Only
Protein Aliases: fj24f03; NR1A2; Nuclear receptor subfamily 1 group A member 2; Thyroid hormone receptor beta; thyroid hormone receptor beta 2; Thyroid hormone receptor beta-1; TR-beta; TR[b]1; trb; TRbeta1; wu:fj24f03
Gene Aliases: fj24f03; NR1A2; si:ch211-264a6.2; thr2; thrb; TR-beta; TR[b]1; trb; wu:fj24f03
UniProt ID: (Zebrafish) Q9PVE4
Entrez Gene ID: (Zebrafish) 30607
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.