Abnova
Manufacturer Code:H00006901M13
Catalog # 89-000-999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F11 |
Immunogen | TAZ (AAH11515 1 a.a. ~ 262 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant TAZ.
For Research Use Only
Protein Aliases: BTHS; CMD3A; EFE; EFE2; FLJ27390; G4.5; LVNCX; Protein G4.5; Tafazzin; Taz1; XAP-2
Gene Aliases: BTHS; CMD3A; EFE; EFE2; G4.5; LVNCX; TAZ; Taz1
UniProt ID: (Human) Q16635
Entrez Gene ID: (Human) 6901
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.