Invitrogen
Manufacturer Code:MA535462
Catalog # PIMA535462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0928 |
Immunogen | A synthesized peptide derived from human PLAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 3.4.21.68 Alteplase AU020998 AW212668 D8Ertd2e PATISS Plat Reteplase t plasminogen activator Tissue-type plasminogen activator Tissue-type plasminogen activator chain A Tissue-type plasminogen activator chain B tPA t-PA; Alteplase; plasminogen/activator kringle; Reteplase; t-PA; t-plasminogen activator; tissue plasminogen activator; Tissue-type plasminogen activator; Tissue-type plasminogen activator chain A; Tissue-type plasminogen activator chain B
Gene Aliases: AU020998; AW212668; D8Ertd2e; PATISS; PLAT; T-PA; TPA
UniProt ID: (Human) P00750, (Mouse) P11214, (Rat) P19637
Entrez Gene ID: (Human) 5327, (Mouse) 18791, (Rat) 25692
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.