Invitrogen
Manufacturer Code:PA551927
Catalog # PIPA551927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human tPA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alteplase; plasminogen/activator kringle; Reteplase; t-PA; T-PA TPA alteplase plasminogen activator tissue type reteplase t-plasminogen activator tissue plasminogen activator (t-PA) tissue-type plasminogen activator; t-plasminogen activator; Tissue-type plasminogen activator; Tissue-type plasminogen activator chain A; Tissue-type plasminogen activator chain B
Gene Aliases: PLAT; T-PA; TPA
UniProt ID: (Human) P00750
Entrez Gene ID: (Human) 5327
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.