Novus Biologicals
Manufacturer Code:NBP274568B
Catalog # NBP274568B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3H3 |
Immunogen | Recombinant protein fragment corresponding to amino acids 157-426 of human t-Plasminogen Activator/tPA (NP_000921) produced in E.coli. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Alteplase; Alteplase DKFZp686I03148 EC 3.4.21 EC 3.4.21.68 plasminogen activator tissue plasminogen activator tissue type reteplase tissue plasminogen activator (t-PA) tissue-type plasminogen activator TPA T-PA t-plasminogen activator; plasminogen/activator kringle; Reteplase; t-PA; t-plasminogen activator; Tissue-type plasminogen activator; Tissue-type plasminogen activator chain A; Tissue-type plasminogen activator chain B
Gene Aliases: PLAT; T-PA; TPA
UniProt ID: (Human) P00750
Entrez Gene ID: (Human) 5327
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.