Abnova
Manufacturer Code:H00006810A01
Catalog # 89-004-328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | STX4A (NP_004595 19 a.a. ~ 119 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant STX4A.
For Research Use Only
Protein Aliases: p35-2; Renal carcinoma antigen NY-REN-31; STX4A; syntaxin 4A (placental); Syntaxin-4
Gene Aliases: p35-2; STX4; STX4A
UniProt ID: (Human) Q12846
Entrez Gene ID: (Human) 6810
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.