Abnova
Manufacturer Code:H00006804M02
Catalog # 89-000-991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B11-1A8 |
Immunogen | STX1A (AAH03011.1 1 a.a. ~ 251 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant STX1A.
For Research Use Only
Protein Aliases: HPC-1; Neuron-specific antigen HPC-1; p35-1; STX1; syntaxin 1A (brain); Syntaxin-1A
Gene Aliases: HPC-1; P35-1; STX1; STX1A; SYN1A
UniProt ID: (Human) Q16623
Entrez Gene ID: (Human) 6804
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.