Abnova
Manufacturer Code:H00008871D01P
Catalog # 89-018-618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SYNJ2 (AAH43277.1 1 a.a. ~ 839 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human SYNJ2 protein.
For Research Use Only
Protein Aliases: inositol phosphate 5'-phosphatase 2; inositol polyphosphate-5-phosphatase H; INPP5H; KIAA0348; MGC44422; phosphoinositide 5-phosphatase; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 2; Synaptojanin-2
Gene Aliases: INPP5H; KIAA0348; SYNJ2
UniProt ID: (Human) O15056
Entrez Gene ID: (Human) 8871
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.