Abnova
Manufacturer Code:H00006476A01
Catalog # 89-018-840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SI (NP_001032 1728 a.a. ~ 1826 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SI.
For Research Use Only
Protein Aliases: alpha-glucosidase; Isomaltase; MGC131621; MGC131622; oligosaccharide alpha-1,6-glucosidase; Sucrase; sucrase-isomaltase (alpha-glucosidase); Sucrase-isomaltase, intestinal
Gene Aliases: SI
UniProt ID: (Human) P14410
Entrez Gene ID: (Human) 6476
Molecular Function:
glucosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.