Abnova
Manufacturer Code:H00008987M03
Catalog # 89-001-110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H4 |
Immunogen | GENX-3414 (AAH22301 1 a.a. ~ 358 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant GENX-3414.
For Research Use Only
Protein Aliases: FLJ41801; genethonin 1; Genethonin-1; GENEX3414; GENX-3414; Glycophagy cargo receptor STBD1; Starch-binding domain-containing protein 1
Gene Aliases: GENEX3414; GENX-3414; STBD1
UniProt ID: (Human) O95210
Entrez Gene ID: (Human) 8987
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.