Abnova
Manufacturer Code:PAB11586
Catalog # 89-115-200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to zebrafish sox9a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of sox9a.
For Research Use Only
Protein Aliases: jef; jellyfish; SRY (sex determining region Y)-box 9a; SRY-box containing gene 9a; wu:fj17b12; zgc:111921
Gene Aliases: jef; wu:fj17b12; zgc:111921
Entrez Gene ID: (Zebrafish) 60641
Molecular Function: HMG box transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.