Abnova
Manufacturer Code:H00006578A01
Catalog # 89-004-313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLCO2A1 (NP_005621 276 a.a. ~ 325 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SLCO2A1.
For Research Use Only
Protein Aliases: matrin F/G 1; OATP2A1; PGT; Prostaglandin transporter; SLC21A2; solute carrier family 21 (prostaglandin transporter), member 2; Solute carrier family 21 member 2; Solute carrier organic anion transporter family member 2A1; solute carrier organic anion transporter family, member 2A1
Gene Aliases: MATR1; OATP2A1; PGT; PHOAR2; SLC21A2; SLCO2A1
UniProt ID: (Human) Q92959
Entrez Gene ID: (Human) 6578
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.