Abnova
Manufacturer Code:H00006520D01P
Catalog # 89-018-269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC3A2 (NP_001013269.1 1 a.a. ~ 529 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human SLC3A2 protein.
For Research Use Only
Protein Aliases: 4F2; 4F2 cell-surface antigen heavy chain; 4F2 heavy chain antigen; 4F2HC; 4T2HC; antigen defined by monoclonal antibody 4F2, heavy chain; antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43; CD98; CD98 heavy chain; CD98HC; Lymphocyte activation antigen 4F2 large subunit; MDU1; monoclonal antibody 44D7; NACAE; solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2; solute carrier family 3 (amino acid transporter heavy chain), member 2; Solute carrier family 3 member 2
Gene Aliases: 4F2; 4F2HC; 4T2HC; CD98; CD98HC; MDU1; NACAE; SLC3A2
UniProt ID: (Human) P08195
Entrez Gene ID: (Human) 6520
Molecular Function:
amylase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.