Abnova
Manufacturer Code:H00005250B01P
Catalog # 89-016-880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC25A3 (AAH00998 1 a.a. ~ 361 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SLC25A3 protein.
For Research Use Only
Protein Aliases: OK/SW-cl.48; PHC; Phosphate carrier protein, mitochondrial; Phosphate transport protein; PTP; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 3; Solute carrier family 25 member 3
Gene Aliases: OK/SW-cl.48; PHC; PTP; SLC25A3
UniProt ID: (Human) Q00325
Entrez Gene ID: (Human) 5250
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.