Abnova
Manufacturer Code:H00055186B01
Catalog # 89-007-813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC25A36 (AAH14064 1 a.a. ~ 311 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SLC25A36 protein.
For Research Use Only
Protein Aliases: DKFZp564C053; FLJ10618; solute carrier family 25 (pyrimidine nucleotide carrier ), member 36; solute carrier family 25 (pyrimidine nucleotide carrier), member 36; Solute carrier family 25 member 36; solute carrier family 25, member 36
Gene Aliases: PNC2; SLC25A36
UniProt ID: (Human) Q96CQ1
Entrez Gene ID: (Human) 55186
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.