Abnova
Manufacturer Code:H00010786B01
Catalog # 89-007-056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC17A3 (AAH17952.1 1 a.a. ~ 420 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SLC17A3 protein.
For Research Use Only
Protein Aliases: Na(+)/PI cotransporter 4; NPT4; Sodium-dependent phosphate transport protein 4; Sodium/phosphate cotransporter 4; solute carrier family 17 (organic anion transporter), member 3; solute carrier family 17 (sodium phosphate), member 3; Solute carrier family 17 member 3
Gene Aliases: GOUT4; NPT4; SLC17A3; UAQTL4
UniProt ID: (Human) O00476
Entrez Gene ID: (Human) 10786
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.