Abnova
Manufacturer Code:H00006563B01P
Catalog # 89-016-906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC14A1 (AAH50539 1 a.a. ~ 389 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SLC14A1 protein.
For Research Use Only
Protein Aliases: FLJ33745; FLJ41687; HsT1341; HUT11; JK; Kidd blood group; RACH1; SLC14A1 JK; solute carrier family 14 (urea transporter), member 1 (Kidd blood group); Solute carrier family 14 member 1; Urea transporter 1; urea transporter JK glycoprotein; Urea transporter, erythrocyte; urea transporter-B1; UT-B1; UT1; UTE
Gene Aliases: HsT1341; HUT11; JK; RACH1; RACH2; SLC14A1; UT-B1; UT1; UTE
UniProt ID: (Human) B3KR62
Entrez Gene ID: (Human) 6563
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.