Abnova
Manufacturer Code:H00026228D01P
Catalog # 89-018-127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STAP1 (NP_036240.1 1 a.a. ~ 295 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human STAP1 protein.
For Research Use Only
Protein Aliases: BCR downstream-signaling protein 1; BRDG1; Docking protein BRDG1; Signal-transducing adaptor protein 1; STAP-1; Stem cell adaptor protein 1
Gene Aliases: BRDG1; STAP-1; STAP1
UniProt ID: (Human) Q9ULZ2
Entrez Gene ID: (Human) 26228
Molecular Function:
protein kinase receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.