Abnova
Manufacturer Code:H00006722M03
Catalog # 89-000-980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E1 |
Immunogen | SRF (AAH48211 406 a.a. ~ 508 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SRF.
For Research Use Only
Protein Aliases: c-fos serum response element-binding transcription factor; MCM1; minichromosome maintenance 1 homolog; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); SRF
Gene Aliases: MCM1; SRF
UniProt ID: (Human) P11831
Entrez Gene ID: (Human) 6722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.