Abnova
Manufacturer Code:H00006690M01
Catalog # 89-014-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D4 |
Immunogen | SPINK1 (AAH25790 24 a.a. ~ 79 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SPINK1.
For Research Use Only
Protein Aliases: Pancreatic secretory trypsin inhibitor; PCTT; PSTI; Serine protease inhibitor Kazal-type 1; serine protease inhibitor, Kazal type 1; Spink3; TATI; Tumor-associated trypsin inhibitor
Gene Aliases: PCTT; PSTI; SPINK1; Spink3; TATI; TCP
UniProt ID: (Human) P00995
Entrez Gene ID: (Human) 6690
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.