Abnova
Manufacturer Code:PAB8990
Catalog # 89-134-075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to fruit fly sad. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of sad.
For Research Use Only
Protein Aliases: 315a1; CG14728; CG14728-PA; CYP315A1; CYPCCCXVA1; cytochrome P450; Cytochrome P450 315a1, mitochondrial; ecdysteroid hydroxylase; karmoisin ghost; LP13573p; Protein shadow; sad-PA
Gene Aliases: 315a1; CG14728; CYP315A1; Dmel\CG14728; Dmel_CG14728; Sad
UniProt ID: (Fruit fly) Q9VGH1
Entrez Gene ID: (Fruit fly) 44858
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.