Novus Biologicals
Manufacturer Code:NBP298217AF350
Catalog # NB081375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant sFRP-4 (NP_057896.1 Met 1-Ser 351). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Frizzled protein human endometrium FRP-4 frpHE FRPHEMGC26498 secreted frizzled-related protein 4 secreted frizzled-related protein 4 secreted frizzled-related protein 4 sFRP-4; Frizzled protein, human endometrium; FrpHE; Secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; sFRP-4
Gene Aliases: FRP-4; FRPHE; sFRP-4; SFRP4
UniProt ID: (Human) Q6FHJ7
Entrez Gene ID: (Human) 6424
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.