Novus Biologicals
Manufacturer Code:NBP211898FR
Catalog # NBP211898FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0546-9J18 |
Immunogen | Human FRZB |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FIZ FREOS1 Frezzled Fritz frizzled-related protein Frizzled-related protein 1 FRP FRP-3 FrzB-1 FRZB1sFRP-3 FRZB-PEN FZRB hFIZ secreted frizzled-related protein 3 SFRP3frezzled SRFP3frizzled homolog-related; Frezzled; Fritz; frizzled homolog-related; Frizzled-related protein 1; FrzB-1; Secreted frizzled-related protein 3; sFRP-3
Gene Aliases: FIZ; FRE; FRITZ; FRP; FRP-3; FRZB; FRZB-1; FRZB-PEN; FRZB1; FZRB; hFIZ; OS1; SFRP3; SRFP3
UniProt ID: (Human) Q92765
Entrez Gene ID: (Human) 2487
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.