R&D Systems
Manufacturer Code:AF1384
Catalog # AF1384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human sFRP-1 (R&D Systems Catalog # 1384-SF) Ser32-Lys314 Accession # AAB70793 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: frizzled-related protein; FRP-1; FRP1 FRP-1FrzA FRPSARP-2 FrzA SARP-2 SARP2sFRP-1 Secreted apoptosis-related protein 2 secreted frizzled-related protein 1 sFRP1; SARP-2; Secreted apoptosis-related protein 2; Secreted frizzled-related protein 1; sFRP-1
Gene Aliases: FRP; FRP-1; FRP1; FrzA; SARP2; SFRP1
UniProt ID: (Human) Q8N474
Entrez Gene ID: (Human) 6422
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.