Abnova
Manufacturer Code:H00051444D01P
Catalog # 89-018-481
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNF138 (NP_057355.2 1 a.a. ~ 245 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human RNF138 protein.
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase RNF138; HSD-4; MGC8758; NARF; nemo-like kinase associated ring finger protein; Nemo-like kinase-associated RING finger protein; NLK-associated RING finger protein; RING finger protein 138; ring finger protein 138, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF138; STRIN; testicular tissue protein Li 165
Gene Aliases: hNARF; HSD-4; HSD4; NARF; RNF138; STRIN
UniProt ID: (Human) Q9UF87
Entrez Gene ID: (Human) 51444
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.