Abnova
Manufacturer Code:H00051109M01
Catalog # 89-014-310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H6 |
Immunogen | RDH11 (AAH00112 24 a.a. ~ 318 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant RDH11.
For Research Use Only
Protein Aliases: Androgen-regulated short-chain dehydrogenase/reductase 1; ARSDR1; CGI-82; FLJ32633; HCBP12; HCV core-binding protein HCBP12; MDT1; prostate short-chain dehydrogenase reductase 1; Prostate short-chain dehydrogenase/reductase 1; PSDR1; RALR1; Retinal reductase 1; Retinol dehydrogenase 11; SCALD; SDR7C1; Short chain dehydrogenase/reductase family 7C member 1; short chain dehydrogenase/reductase family 7C, member 1
Gene Aliases: ARSDR1; CGI-82; CGI82; HCBP12; MDT1; PSDR1; RALR1; RDH11; RDJCSS; SCALD; SDR7C1
UniProt ID: (Human) Q8TC12
Entrez Gene ID: (Human) 51109
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.