Abnova
Manufacturer Code:PAB1172
Catalog # 89-115-325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to full length <i>Schizosaccharomyces pombe</i> rad51. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against full-length recombinant Schizosaccharomyces pombe rad51.
For Research Use Only
Protein Aliases: DNA repair protein rhp51; RAD51 homolog; rhp51
Gene Aliases: rad51; rhp51; SPAC644.14c
UniProt ID: (Fission yeast) P36601
Entrez Gene ID: (Fission yeast) 2543580
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.