Abnova
Manufacturer Code:PAB1173
Catalog # 89-115-336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to full length <i>Schizosaccharomyces pombe</i> rad22. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyclonal antibody raised against full-length recombinant Schizosaccharomyces pombe rad22.
For Research Use Only
Protein Aliases: DNA repair and recombination protein rad22
Gene Aliases: rad22; rad22A; SPAC30D11.10
UniProt ID: (Fission yeast) P36592
Entrez Gene ID: (Fission yeast) 2543086
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.