Abnova
Manufacturer Code:H00005860M01
Catalog # 89-000-926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D4-1D3 |
Immunogen | QDPR (AAH00576 1 a.a. ~ 244 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant QDPR.
For Research Use Only
Protein Aliases: 6,7-dihydropteridine reductase; DHPR; Dihydropteridine reductase; FLJ42391; HDHPR; PKU2; Quinoid dihydropteridine reductase; SDR33C1; Short chain dehydrogenase/reductase family 33C member 1; short chain dehydrogenase/reductase family 33C, member 1; testis secretory sperm-binding protein Li 236P
Gene Aliases: DHPR; PKU2; QDPR; SDR33C1
UniProt ID: (Human) P09417
Entrez Gene ID: (Human) 5860
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.