Abnova
Manufacturer Code:H00005164M02
Catalog # 89-000-872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5F8 |
Immunogen | PDK2 (AAH05811 187 a.a. ~ 276 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant PDK2.
For Research Use Only
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2, mitochondrial; PDH kinase 2; PDHK2; Pyruvate dehydrogenase kinase isoform 2; pyruvate dehydrogenase kinase, isoenzyme 2; pyruvate dehydrogenase, lipoamide, kinase isozyme 2, mitochondrial
Gene Aliases: PDHK2; PDK2; PDKII
UniProt ID: (Human) Q15119
Entrez Gene ID: (Human) 5164
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.