Abnova
Manufacturer Code:H00008566A01
Catalog # 89-004-397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDXK (NP_003672 51 a.a. ~ 160 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PDXK.
For Research Use Only
Protein Aliases: C21orf124; C21orf97; DKFZp566A071; epididymis secretory sperm binding protein Li 1a; FLJ31940; FLJ37311; MGC15873; MGC31754; MGC52346; PKH; PNK; PRED79; Pyridoxal kinase; pyridoxamine kinase; Pyridoxine kinase; vitamin B6 kinase
Gene Aliases: C21orf124; C21orf97; HEL-S-1a; PDXK; PKH; PNK; PRED79
UniProt ID: (Human) O00764
Entrez Gene ID: (Human) 8566
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.