Abnova
Manufacturer Code:H00008575M01
Catalog # 89-014-223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B9-1A7 |
Immunogen | PRKRA (AAH09470 1 a.a. ~ 313 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant PRKRA.
For Research Use Only
Protein Aliases: DYT16; HSD14; interferon-inducible double stranded RNA-dependent protein kinase activator A; Interferon-inducible double-stranded RNA-dependent protein kinase activator A; PACT; PKR-associated protein X; PKR-associating protein X; Protein activator of the interferon-induced protein kinase; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; Protein kinase, interferon-inducible double-stranded RNA-dependent activator; RAX
Gene Aliases: DYT16; HSD-14; HSD14; PACT; PRKRA; RAX
UniProt ID: (Human) O75569
Entrez Gene ID: (Human) 8575
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.