Abnova
Manufacturer Code:H00005721M02
Catalog # 89-014-175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G4 |
Immunogen | PSME2 (NP_002809 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant PSME2.
For Research Use Only
Protein Aliases: 11S regulator complex beta subunit; 11S regulator complex subunit beta; Activator of multicatalytic protease subunit 2; cell migration-inducing protein 22; MCP activator, 31-kD subunit; PA28B; PA28beta; proteasome (prosome, macropain) activator subunit 2 (PA28 beta); Proteasome activator 28 subunit beta; proteasome activator 28-beta; Proteasome activator complex subunit 2; proteasome activator hPA28 subunit beta; REG-beta; REGbeta
Gene Aliases: PA28B; PA28beta; PSME2; REGbeta
UniProt ID: (Human) Q9UL46
Entrez Gene ID: (Human) 5721
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.