Invitrogen
Manufacturer Code:PA5111806
Catalog # PIPA5111806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 84-104 of rat NGF (precursor) (Pro-domain of the NGF protein) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: B nerve growth factor Beta-nerve growth factor BetaNGF Beta-NGF bNGF HSAN5 nerve growth factor NGF NGF b NGF beta NGF NGFB NGFbeta proNGF nerve growth factor NGF NGF; Beta-nerve growth factor; Beta-NGF; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta subunit
Gene Aliases: Beta-NGF; HSAN5; NGF; NGFB
UniProt ID: (Human) P01138, (Mouse) P01139, (Rat) P25427
Entrez Gene ID: (Human) 4803, (Mouse) 18049, (Rat) 310738
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.