Novus Biologicals
Manufacturer Code:NBP275627
Catalog # NBP275627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM08-36 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Alpha-atrial natriuretic peptide; Alpha-hANP; ANF; ANF 1-33; ANF 3-33; ANF 8-33; ANF ANPatriopeptin ATFB6 atrial natriuretic factor Atrial natriuretic peptide CDD-ANF natriuretic peptide A natriuretic peptide precursor A PNDcardionatrin prepronatriodilatin; ANP; Atrial natriuretic factor; Atrial natriuretic factor 1-33; Atrial natriuretic factor 3-33; Atrial natriuretic factor 8-33; Atrial natriuretic factor prohormone; Atrial natriuretic peptide; Atrial natriuretic peptide prohormone; Atriopeptigen; atriopeptin; Atriopeptin I; Atriopeptin II; Atriopeptin III; Atriopeptin-1; Atriopeptin-2; Atriopeptin-3; Auriculin-A; Auriculin-B; Auriculin-C; Auriculin-D; Cardiodilatin; cardiodilatin-related peptide; Cardionatrin; CDD; CDD 95-126; CDD-ANF; CDD-ANP (95-126); CDD-ANP (99-126); Kaliuretic peptide; KP; LANH; LANP; Long-acting natriuretic hormone; Long-acting natriuretic peptide; natriuretic peptide precursor A variant 1; Natriuretic peptides A; preproANP; preproCDD-ANF; prepronatriodilatin; Pro atrial natriuretic factor 1-30; Pro atrial natriuretic factor 31-67; Pro atrial natriuretic factor 79-98; Pro atrial natriuretic factor 99-126; Pro atrial natriuretic peptide 1-30; Pro atrial natriuretic peptide 31-67; Pro atrial natriuretic peptide 79-98; Pro atrial natriuretic peptide 95-126; proANF; proANF 1-30; proANF 31-67; proANF 79-98; proANF 99-126; proANP 1-30; proANP 31-67; proANP 79-98; proANP 95-126; URO; Urodilatin; Vessel dilator; VSDL
Gene Aliases: ANF; ANP; ATFB6; ATRST2; CDD; CDD-ANF; CDP; NPPA; PND
UniProt ID: (Human) P01160
Entrez Gene ID: (Human) 4878
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.