Abnova
Manufacturer Code:H00005090M02
Catalog # 89-000-863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A11 |
Immunogen | PBX3 (NP_006186 342 a.a. ~ 434 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant PBX3.
For Research Use Only
Protein Aliases: Homeobox protein PBX3; pre-B-cell leukemia homeobox 3; Pre-B-cell leukemia transcription factor 3
Gene Aliases: PBX3
UniProt ID: (Human) P40426
Entrez Gene ID: (Human) 5090
Molecular Function: DNA-directed RNA polymerase RNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.