Abnova
Manufacturer Code:H00003753M13
Catalog # 89-000-766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A6 |
Immunogen | KCNE1 (AAH36452 1 a.a. ~ 105 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant KCNE1.
For Research Use Only
Protein Aliases: cardiac delayed rectifier potassium channel protein; Delayed rectifier potassium channel subunit IsK; FLJ18426; FLJ38123; FLJ94103; IKs producing slow voltage-gated potassium channel subunit beta Mink; ISK; JLNS; JLNS2; LQT2/5; LQT5; MGC33114; Minimal potassium channel; MinK; potassium channel, voltage gated subfamily E regulatory beta subunit 1; Potassium voltage-gated channel subfamily E member 1; potassium voltage-gated channel, Isk-related family, member 1; potassium voltage-gated channel, Isk-related subfamily, member 1; voltage gated potassiun channel accessory subunit
Gene Aliases: ISK; JLNS; JLNS2; KCNE1; LQT2/5; LQT5; MinK
UniProt ID: (Human) P15382
Entrez Gene ID: (Human) 3753
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.