Abnova
Manufacturer Code:H00011232B01P
Catalog # 89-017-021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | POLG2 (AAH09194.1 1 a.a. ~ 485 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human POLG2 protein.
For Research Use Only
Protein Aliases: DNA polymerase gamma accessory 55 kDa subunit; DNA polymerase subunit gamma-2, mitochondrial; HP55; Mitochondrial DNA polymerase accessory subunit; mitochondrial DNA polymerase subunit gamma-2; mitochondrial DNA polymerase, accessory subunit; MTPOLB; p55; PEOA4; POLB; POLG-BETA; POLGB; polymerase (DNA directed), gamma 2, accessory subunit; polymerase (DNA) gamma 2, accessory subunit
Gene Aliases: HP55; MTPOLB; PEOA4; POLB; POLG-BETA; POLG2; POLGB
UniProt ID: (Human) Q9UHN1
Entrez Gene ID: (Human) 11232
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.