Abnova
Manufacturer Code:H00005326A01
Catalog # 89-004-243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLAGL2 (NP_002648 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PLAGL2.
For Research Use Only
Protein Aliases: C2H2-type zinc finger protein; FLJ23283; pleiomorphic adenoma gene-like 2; Pleiomorphic adenoma-like protein 2; Zinc finger protein PLAGL2
Gene Aliases: KIAA0198; PLAGL2; ZNF900
UniProt ID: (Human) Q9UPG8
Entrez Gene ID: (Human) 5326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.